Principaux documents
Z660469
Julabo® bath test tube trays
holds 360 test tubes
About This Item
Code UNSPSC :
41122804
Nomenclature NACRES :
NB.77
Matériaux
stainless steel
Caractéristiques
Holds 360 x test tubes
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8960441
Disponibilité
available only in EU
Description générale
Carrier trays with 4 racks 8970381 (Z615684-1EA) to hold 360 tubes, tube size 75 x 12/13 mm, suitable for use up to to +80 °C.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.