Principaux documents
Z657182
IKA® A11 basic grinding chambers
A 11.5 grinding chamber, 1/cs
Synonyme(s) :
grinders, grinding chambers, grinding mills
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.74
Caractéristiques
CE compliant
CSA approved
Conditionnement
pack of 1
Fabricant/nom de marque
IKA 0002983100
Taille
80 mL , effective volume
Description générale
Replacement grinding bowl for use with the IKA A11 Basic Analytical Mill. Made of Tefcel (ETFE, glass fiber reinforced) with stainless steel inlet (AISI 316L), usable volume 80ml. Has good chemical and low temperature resistance down to -200 °C.
Informations légales
IKA is a registered trademark of IKA-Werke GmbH & Co. KG