Principaux documents
Z562645
Sigma-Aldrich
Open-edged fritted disc porosity assortment pack for VerSAfunnels
disc diam. 24 mm
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Diam. du disque
24 mm
Description générale
Each pack consists of 1 of each porosity:
4-8μm
10-20μm
25-50μm
70-100μm
145-175μm
The 90mm disc pack does not include the 4-8μm porosity.
Informations légales
VerSAFunnel is a trademark of Sigma-Aldrich Co. LLC