Principaux documents
Z558079
Compresseur à 6 emplacements Whatman®
6-place compressor, pack of 1
Synonyme(s) :
Whatman® 6-place compressor, syringeless filters
About This Item
Code UNSPSC :
41104900
Matériaux
polypropylene housing
Conditionnement
pack of 1
Fabricant/nom de marque
Whatman Article No. 28421456 (US reference)
Whatman CR0000006
Description générale
Station manuelle pour travailler avec 6 filtres sans seringue Mini-Uniprep®en même temps, réduction du stress de manipulation et augmentation du rendement de travail.
Autres remarques
Global SIAL customers (excluding Japan) can now order any product from the Cytiva Life Sciences consumables portfolio (prepack only) using GE′s product numbering system.
The legacy GE Whatman products that use the SIAL "Z" product numbering system will be discontinued and directly replaced with a product number using a "WHA" prefix plus the GE product code. For reference, the legacy "Z" product number is identified as a synonym on the "WHA" product number listing.
Informations légales
Mini-UniPrep is a registered trademark of Cytiva
Uniprep is a registered trademark of Whatman International Ltd.
Whatman is a registered trademark of Cytiva