Principaux documents
Z557862
Sigma-Aldrich
Aldrich® solvent storage/dispensing flask, graduated, w/ Sure/Stor inlet
top joint: ST/NS 24/40, bottom joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
Joint
bottom joint: ST/NS14/20
top joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
200 mL
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC