Principaux documents
Z555126
Sigma-Aldrich
Open-edged fritted disc for Aldrich® Buchner VerSAfunnels
disc diam. 50 mm, porosity 145-175 μm
About This Item
Code UNSPSC :
41121809
Diam. du disque
50 mm
Épaisseur
7.5 mm
Dimension de pores
145-175 μm porosity
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
VerSAFunnel is a trademark of Sigma-Aldrich Co. LLC