Principaux documents
Z555029
Sigma-Aldrich
Open-edged fritted disc for Aldrich® Buchner VerSAfunnels
disc diam. 24 mm, porosity 10-20 μm
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Diam. du disque
24 mm
Épaisseur
7.5 mm
Dimension de pores
10-20 μm porosity
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
VerSAFunnel is a trademark of Sigma-Aldrich Co. LLC