Z549789
Aldrich® jacketed three-neck flask
half jacket, capacity 500 mL, center joint: ST/NS 24/40
Synonyme(s) :
boiling flask, flask round, flask round bottom
2 460,00 $
About This Item
2 460,00 $
Produits recommandés
Matériaux
borosilicate glass
round bottom
three-neck
three-neck flask
Joint
center joint: ST/NS24/40
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
500 mL
Catégories apparentées
Description générale
Features:
- Eliminates dead space for complete stirring of flask contents
- Allows for complete draining without breaking down flask set-up
- Liquid cooled or heated with liquid or steam
- 24/40 side joints