Z549606
Stopcock Adapter
PTFE side-port stopcock, joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Matériaux
PTFE side-port stopcock
Joint
joint: ST/NS14/20
Catégories apparentées
Description générale
Glass and PTFE stopcocks have a 2 mm bore and accommodate 1/4 in. I.D. tubing. Can be used in vacuum or atmospheric applications.