Documents
Z547735
Sigma-Aldrich
Aldrich® Buchner funnel with joint
capacity 30 mL, disc diam. 30 mm, porosity 10-20 μm, joint: ST/NS 14/20
Synonyme(s) :
buchner funnels, filter funnel, filter funnels
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/20
Capacité
30 mL
Diam. du disque
30 mm
Dimension de pores
10-20 μm porosity
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC