MilliporeSigma
Toutes les photos(6)
Principaux documents
Z545465
Molymod® Individual organic model set
Protein (beta pleated sheet)
1 SET
81,20 $
81,20 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 SET
81,20 $
About This Item
Code UNSPSC :
60104506
Nomenclature NACRES :
NB.17
81,20 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Description
peptides units: 15
Fabricant/nom de marque
Molymod
Description générale
The mushroom-shaped hydrogen Atomlink components and the short (non-visible) bonding links provide compact, semi-space-filling models with an internuclear scale of 1.45 cm per Angstrom. Polymer models include long, flexible links for demonstration of the mechanism of polymerization.
Informations légales
Atomlink is a trademark of Spiring Enterprises Ltd.
Molymod is a registered trademark of Spiring Enterprises Ltd.