Principaux documents
Z544523
Molymod® molecular model set
Student Organic/Inorganic
1 SET
55,90 $
55,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 SET
55,90 $
About This Item
Code UNSPSC :
60104506
Nomenclature NACRES :
NB.17
55,90 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
no. of atoms 52
no. of bonds 38
Fabricant/nom de marque
Molymod MMS-009
Description générale
14 hydrogen (l), 6 carbon (lV), 7 oxygen (ll, lV), 3 nitrogen (lll, lV), 2 sulphur (ll, Vl), 2 phosphorus (lll, V), 6 halogen (l), 8 metal (l, ll, lll, lV, Vl), 4 brown (lll, lV, V, Vl), 3 lone pair 2D paddles (beige) Links: 20 medium (grey), 5 medium (purple), 12 long flexible (grey)
Top quality Molymod molecular model sets are for use by students, teachers, and researchers. May be upgraded or customized with additional model parts, for more info, go to sigma-aldrich.com and enter Molymod in our search engine.
Scale is 2.8 cm =100 pm (open style) and 1.45 cm = 100 pm (compact, semi-space-filling). Sets include standard color-coded plastic atom centers (with bond holes set at the appropriate angles), bonding links, and a plastic storage box.
Set pictured is representative of the Student set.
See our Selection Guide for detailed descriptions of set contents.
Informations légales
Molymod is a registered trademark of Spiring Enterprises Ltd.