Principaux documents
Z531456
Sigma-Aldrich
Aldrich® Graham condenser
Overall H 400 mm, joint: ST/NS 29/32
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/32
Disponibilité
not available in EU
H totale
400 mm
Description générale
The Graham condenser has a large cooling surface area by allowing coolant to circulate around the inner coil. Condensers have a standard taper outer joint and a lower inner drip-tip joint.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC