Principaux documents
Z516678
Sigma-Aldrich
Aldrich® three-neck round-bottom flask
angled, capacity 500 mL, joint: ST/NS 34/45, side Joints: ST/NS 24/40 (2)
Synonyme(s) :
flask round bottom, round bottom flasks, round bottomed flask
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
three angled neck
three-neck
Joint
side Joints: ST/NS24/40 (2)
joint: ST/NS34/45
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Sigma-Aldrich
Capacité
500 mL
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC