Z502413
Glindemann PTFE sealing rings
(for 29/26, 29/42, 29/32 joints)
Synonyme(s) :
ptfe ring, ptfe sealing
About This Item
Produits recommandés
Joint
joint 29/26
joint 29/32
joint 29/42
Conditionnement
pkg of 50 ea
pkg of
Fabricant/nom de marque
Glindemann K29
Plage de mesure de la température
≤250 °C
Description générale
- This narrow ring seal is an alternative to a PTFE joint sleeve
- Greaseless protection for air- and moisture- sensitive compounds
- Can be used for prolonged storage under vacuum
- Slide easily onto medium and full length taper joints
- Can be used with Keck® clip and similar metal joint clips