Z414123
Aldrich® offset enlarging adapter
male joint: ST/NS 24/40, female joint: ST/NS 34/45
Synonyme(s) :
glass adapter, glass adapters
About This Item
Joint
female joint: ST/NS34/45
male joint: ST/NS24/40
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC