Principaux documents
Z309281
Quickfit® flat-bottom boiling flasks
capacity 100 mL, joint: ST/NS 19/26
Synonyme(s) :
FF100/2S
About This Item
Code UNSPSC :
41121804
Matériaux
borosilicate glass
flat bottom flask
short
Joint
joint: ST/NS19/26
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 5
Fabricant/nom de marque
Quickfit FF 100/2S
Disponibilité
not available in USA and Canada
Capacité
100 mL
Description générale
With short neck and ground-glass socket. Not suitable for vacuum use.
Caractéristiques et avantages
  • Complies with ISO 4797 and ISO 1773
  • Manufactured from 3.3 expansion borosilicate glass complying with ISO 3585 (ASTM E-438 Type 1 Class A)
  • The flat base allows use with hotplates
  • FF150/3S, FF150/4S, FF150/5S and FF500/2S are supplementary to ISO 4797.
  • Note: These flasks are not suitable for use under a positive or negative pressure
Informations légales
Quickfit is a registered trademark of SciLabware Limited