Principaux documents
Z309222
Quickfit® Allihn condenser
L 250 mm, joint: ST/NS 35/35
Synonyme(s) :
CX7/05/SC
About This Item
Code UNSPSC :
41121800
Joint
joint: ST/NS35/35
Fabricant/nom de marque
Quickfit CX 7/05/SC
Disponibilité
not available in USA and Canada
Longueur
250 mm
Superficie
130 cm2
Description générale
Bulb sections give greater surface area for more cooling than the corresponding Liebig type condenser. Generally used for reflux. Condensers have plain stem tops.
Informations légales
Quickfit is a registered trademark of SciLabware Limited