Principaux documents
Z308854
Quickfit® conical flasks
capacity 250 mL, joint: ST/NS 34/35
Synonyme(s) :
FE250/5
About This Item
Code UNSPSC :
41121804
Matériaux
borosilicate glass
conical flask
Joint
joint: ST/NS34/35
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
case of 5
Fabricant/nom de marque
Quickfit FE250/5
Disponibilité
not available in USA and Canada
Capacité
250 mL
Description générale
With ground-glass socket for general use. Not suitable for vacuum use.
Informations légales
Quickfit is a registered trademark of SciLabware Limited