Documents
Z304778
Quickfit® powder funnels
joint: ST/NS 14/23
Synonyme(s) :
CF5/1
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS14/23
Fabricant/nom de marque
Quickfit CF 5/1
Disponibilité
not available in USA and Canada
Catégories apparentées
Description générale
For pouring powders or liquids into vessels with ground-glass joints
Informations légales
Quickfit is a registered trademark of SciLabware Limited