Documents
Z304522
Quickfit® pressure equalising dropping funnel
capacity 250 mL, Pear-shaped, male joint: ST/NS 24/29, female joint: ST/NS 24/29
Synonyme(s) :
DE250/33
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Joint
female joint: ST/NS24/29
male joint: ST/NS24/29
Caractéristiques
Pear-shaped
Fabricant/nom de marque
Quickfit DE 250/33
Disponibilité
not available in USA and Canada
Capacité
250 mL
Catégories apparentées
Description générale
With GP Rotaflo stopcocks
Informations légales
Quickfit is a registered trademark of SciLabware Limited