Principaux documents
Z302376
Quickfit® Cone/Screw-thread Adapters
13 thread (couples 6.0 - 7.0 mm diameter), joint: ST/NS 34/35
Synonyme(s) :
ST55/13
About This Item
Code UNSPSC :
41121815
Joint
joint: ST/NS34/35
Fabricant/nom de marque
Quickfit ST 55/13
Disponibilité
not available in USA and Canada
not available in USA and Canada
Raccords
13 thread (couples 6.0 - 7.0 mm diameter)
Description générale
The Quickfit screw-thread joints on these cone adapters provide a versatile means of coupling thermometers, bleed tubes, electrodes, etc; with Quickfit glassware. The joints are readily adjustable for depth of immersion by slackening the screw cap, and give excellent results under vacuum.
Can be used in apparatus at temperatures up to 300°C providing excessive heat is not applied directly to the screw-thread union.
For replacement parts, see related products section of web site listing.
Informations légales
Quickfit is a registered trademark of SciLabware Limited