Principaux documents
Z301825
Marigold® Suregrip gloves
size L
About This Item
Code UNSPSC :
12352200
Matériaux
yellow natural rubber
Taille
L
Caractéristiques
CE compliant
Conditionnement
pkg of 12 pair
Fabricant/nom de marque
Marigold
Disponibilité
not available in USA and Canada
L × épaisseur
320 mm × 1.50 mm
Description générale
Easy pull-on, tear resistant flock lined yellow rubber gloves offering comfort with a high resistance to water based chemicals. Unique raised pattern gives these gloves an excellent wet grip.
Informations légales
Marigold is a registered trademark of Comasec SAS