Principaux documents
Z301175
Quickfit® expansion adapter
top female joint: ST/NS 34/35, bottom male joint: ST/NS 29/32
Synonyme(s) :
XA54
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Joint
female joint: ST/NS top34/35
male joint: ST/NS bottom29/32
Fabricant/nom de marque
Quickfit XA 54
Disponibilité
not available in USA and Canada
not available in USA and Canada
Description générale
These adapters connect items with larger cone to a piece of glassware with a smaller size socket.
Informations légales
Quickfit is a registered trademark of SciLabware Limited