Z244112
Sample cylinder plug
brass
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
Produits recommandés
Matériaux
brass
Description générale
The 1/4 in. NPTM square head plug fits all Sure/Pac sample cylinder sizes.