Principaux documents
Z236810
Crew® wipers
L × W 11 1/2 in. × 12 in.
About This Item
Code UNSPSC :
41103200
Nomenclature NACRES :
NB.35
Conditionnement
pkg of 100 wipes
Fabricant/nom de marque
Kimberly Clark
L × l
11 1/2 in. × 12 in.
Description générale
Low lint wipers for Class 100 assembly operations and other clean room environments. Soft, absorbent, nonwoven fabric eliminates surface fibers and traps particles. Won′t scratch sensitive surfaces and absorbs more than four times its weight in liquids. Acid resistant-won′t break down or disintegrate when exposed to chemicals. Packaged in anti-static, double polybag.
Informations légales
Crew is a registered trademark of Kimberly-Clark Worldwide, Inc.