Principaux documents
Z179620
Supelco
Michel-Miller end-fitting adapter
Ace-Thred for #7
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.52
Fabricant/nom de marque
Ace Glass 580107
Technique(s)
LPLC: suitable
Raccords
Ace-Thred for #7
Description générale
Use at either end of column. 1/4 -28 female thread at top accepts standard miniature plumbing systems. Male Ace-Thred matches thread size of M-M columns. Bore is 1.5 mm. Use with filter discs to retain column-packing material. Order adapter and filter discs separately.