Principaux documents
Z150215
Sigma-Aldrich
Kilo-Lab® transfer lines
brass valve,
needle valve Luer lock
1 EA
552,00 $
552,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
552,00 $
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.43
552,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
brass valve
Catégories apparentées
Description générale
Convenient fully assembled lines for removing liquid from Kilo-Lab cylinders. Line consists of a brass CGA 510 fitting connected to a 48in. length of metal braided PTFE tubing connected to a metal ball valve with metal needle connector. "T" is placed in-line and is equipped with a needle valve and 6mm I.D. septum inlet for nitrogen flush of transfer line.
Informations légales
Kilo-Lab is a registered trademark of Merck KGaA, Darmstadt, Germany