Principaux documents
Z147540
Supelco
Flow controller for flash-chromatography assembly with threaded joints
joint: ST/NS 45/50
About This Item
Code UNSPSC :
41115710
Nomenclature NACRES :
NC.05
Joint
joint: ST/NS45/50
Qualité
for chromatography
Technique(s)
LPLC: suitable
Adéquation
suitable for chromatography
Liaison
See the Threaded Ground Glass Tutorial to see how threaded joints work.