Principaux documents
Z129259
Sigma-Aldrich
Aldrich® vacuum-filtration adapter
male joint: ST/NS 29/26, flange I.D. 31 mm
About This Item
Code UNSPSC :
41104900
Nomenclature NACRES :
NB.24
Joint
male joint: ST/NS29/26
D.I. du collier
31 mm
Description générale
Enables filtration (via a Buchner funnel) directly into a single-neck, round-bottom flask. Includes rubber sleeve.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC