Documents
Z125377
Sigma-Aldrich
Ace-Thred Thermometer/Tubing Adapter
Straight, joint: ST/NS 19/22, thread I.D. 7.0 mm
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Étiquette/Marqueur
A
Joint
joint: ST/NS19/22
Fabricant/nom de marque
Ace Glass 502828
D.I. du filetage
7.0 mm
Description générale
Threaded nylon bushing with FETFE® O-ring.
Informations légales
Ace-Thred is a trademark of Ace Glass, Inc.
FETFE is a registered trademark of Ace Glass, Inc.
Pièce de rechange