Principaux documents
WHA2105918
Whatman® lens cleaning tissue, Grade 105
W × L 460 mm × 570 mm, pkg of 500 ea
Synonyme(s) :
Z741388, lens cleaning paper, lens paper, lens tissue
1 PKG
622,00 $
622,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
622,00 $
About This Item
Code UNSPSC :
41103200
Nomenclature NACRES :
NB.24
622,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 500 ea
Fabricant/nom de marque
Cytiva
Whatman 2105-918
Whatman Article No. 28418698 (US reference)
Larg. × L
460 mm × 570 mm
Description générale
High-quality whatman lens cleaning tissue, which is chemically pure and free from silicones and other additives. Lenses and other optical surfaces made from glass, quartz or plastic can be easily scratched if you do not clean them with a very soft surface. High quality Whatman lens cleaning tissue provides the solution. The tissue is chemically pure and free from silicones and other additives. Most importantly, it can be relied on to safely remove surface moisture and grease.
Caractéristiques et avantages
  • Soft texture will not damage lenses or optical surfaces.
  • Chemically pure tissue is free from silicones and other additives.
  • High absorbency leads to increased safety upon removal of surface moisture and grease.
  • Very strong and leaves no fibers.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva