Principaux documents
WHA10461100
Whatman® FP 050/0 Filter holder
Synonyme(s) :
Z698512
1 PKG
180,00 $
180,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
180,00 $
About This Item
Code UNSPSC :
41104922
Nomenclature NACRES :
NB.24
180,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Whatman 10461100
Whatman Article No. 28415719 (US reference)
Description générale
The female Luer lock inlet is compatible with typical syringes. Polysulfone housing can be autoclaved with membrane in place for applications requiring a sterile filter assembly.
Caractéristiques et avantages
  • Durable polysulfone plastic construction provides moderate chemical compatibility with lighter weight than stainless steel filter holders.
  • Polysulfone filter holders are often used for clarification, venting applications, particulate removal or collection from aqueous, air or gas streams.
Autres remarques
Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.
Informations légales
Whatman is a registered trademark of Cytiva