Principaux documents
WHA10418412
Whatman® Nuclepore Track-Etched Membranes
polycarbonate filter, filter diam. 47 mm, pore size 10 μm, non-sterile, case of 100 ea
About This Item
Code UNSPSC :
41104921
Nomenclature NACRES :
NB.24
Matériaux
polycarbonate filter
Stérilité
non-sterile
Caractéristiques
hydrophilic
Conditionnement
case of 100 ea
Fabricant/nom de marque
Cytiva 10418412
Diamètre
47 mm
Diam. du filtre
47 mm
Dimension de pores
10 μm
Application
  • Epifluorescence microscopy
  • Environmental analysis
  • Cell biologyEPA testing
  • Fuel testing
  • Bioassays
  • Parasitology
  • Air analysis
  • Water microbiology
Informations légales
Nuclepore is a trademark of Cytiva
Whatman is a registered trademark of Cytiva