Principaux documents
WHA10410014
Whatman® regenerated cellulose RC60 filter discs 1 μm pore size
white, hydrophilic, 100 ea, 50 mm diam
Synonyme(s) :
Z697885, filter membrane, membrane filter, syringe filter
1 PKG
534,00 $
534,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 PKG
534,00 $
About This Item
Code UNSPSC :
40161507
Nomenclature NACRES :
NB.24
534,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Nom du produit
Whatman® regenerated cellulose membrane filters, RC60 Membrane Circles, diam. 50 mm, pore size 1 μm
Caractéristiques
hydrophilic
Conditionnement
pkg of 100 ea
Fabricant/nom de marque
Whatman 10410014
Whatman Article No. 28415270 (US reference)
Diamètre
50 mm
Dimension de pores
1 μm
Description générale
Cytiva′s Whatman regenerated cellulose membrane filters are microporous cellulose acetate filters made without wetting agents. RC60 offers low extractables and wide compatibility in both organic and aqueous media.
Versatile regenerated cellulose syringe filter membranes
Syringe filters require high-quality microporous membrane filters that minimally impact sample analysis with extractable impurities. Regenerated cellulose membrane filters from Cytiva′s business have broad solvent compatibility, and they contribute very low extractable material in a wide variety of sample solvents. Thus, they are appropriate for sample preparation in many applications and as a standalone or syringe filter membrane.
Caractéristiques et avantages
  • Spontaneous wetting with very good wet strength
  • Chemical resistance suitable for a wide range of aqueous and organic media
  • Mechanically stable with low protein binding
  • Low extractable levels to minimize sample contamination
  • Can be sterilized by all common methods
Autres remarques
Field of Use : Any microbiological or biotechnological application, including, but not limited to application of membranes for diagnostic, medical, biological, pharmaceutical, consumer product and food and beverage purposes.
Informations légales
Nitrocellulose Membranes (FFHP, Immunopore and BA membranes)
Sold under license to DE10102744 and foreign equivalents thereof
Whatman is a registered trademark of Cytiva