Principaux documents
W446207
Sigma-Aldrich
Hydroxyacetone
≥95%, FG
Synonyme(s) :
1-Hydroxy-2-propanone, Acetol
About This Item
Formule linéaire :
CH3COCH2OH
Numéro CAS:
Poids moléculaire :
74.08
Numéro FEMA:
4462
Numéro C.I. (Colour Index):
11101
Beilstein:
605368
Numéro CE :
Numéro MDL:
Code UNSPSC :
12164502
ID de substance PubChem :
Numéro Flavis :
7.169
Propriétés organoleptiques:
sweet
Qualité:
FG
Kosher
Source biologique:
synthetic
Le tarif et la disponibilité ne sont pas disponibles actuellement.
Source biologique
synthetic
Niveau de qualité
Qualité
FG
Kosher
Conformité réglementaire
EU Regulation 1334/2008 & 178/2002
Essai
≥95%
Indice de réfraction
n20/D 1.425 (lit.)
pb
145-146 °C (lit.)
Pf
−17 °C (lit.)
Densité
1.082 g/mL at 25 °C (lit.)
Application(s)
flavors and fragrances
Propriétés organoleptiques
sweet
Chaîne SMILES 
CC(=O)CO
InChI
1S/C3H6O2/c1-3(5)2-4/h4H,2H2,1H3
Clé InChI
XLSMFKSTNGKWQX-UHFFFAOYSA-N
Autres remarques
Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.
Pictogrammes
FlameHealth hazard
Mention d'avertissement
Warning
Mentions de danger
Classification des risques
Flam. Liq. 3 - Muta. 2
Code de la classe de stockage
3 - Flammable liquids
Classe de danger pour l'eau (WGK)
WGK 1
Point d'éclair (°F)
133.7 °F - closed cup
Point d'éclair (°C)
56.5 °C - closed cup
Jieni Lian et al.
Bioresource technology, 118, 177-186 (2012-06-19)
The presence of very reactive C1-C4 molecules adversely affects the quality bio-oils produced from the pyrolysis of lignocellulosic materials. In this paper a scheme to produce lipids with Cryptococcus curvatus from the carboxylic acids in the pyrolytic aqueous phase collected
Akshay Kumar et al.
Organic & biomolecular chemistry, 9(8), 2731-2742 (2011-03-02)
A new series of water compatible primary-tertiary diamine catalysts derived from natural primary amino acids bearing a hydrophobic side chain have been synthesized. These new primary-tertiary diamine-Brønsted acid conjugates bifunctional organocatalysts efficiently catalyzes the asymmetric direct syn selective cross-aldol reaction
Maciej Gasior et al.
Epilepsia, 48(4), 793-800 (2007-03-28)
Acetone, one of the principal ketone bodies elevated during treatment with the ketogenic diet, exhibits anticonvulsant properties that may contribute to the seizure protection conferred by the diet. The anticonvulsant mechanism of acetone is unknown, but it is metabolized to
Xianliang Zhou et al.
Environmental science & technology, 43(8), 2753-2759 (2009-05-30)
A method has been modified and optimized for the measurements of hydroxyacetone as well as formaldehyde and glycolaldehyde, based on aqueous scrubbing using a coil sampler followed by DNPH derivatization and HPLC analysis. Derivatization equilibrium and kinetics were studied to
Jacek Mlynarski et al.
Chemical Society reviews, 41(2), 587-596 (2011-07-29)
The key role of carbohydrates in biological processes and their visible existence in our everyday life have stimulated the interest of leading research groups on the smart and simple synthesis of common and rare sugar molecules. Now, more than 120