Documents
TEM-CF400HCU50
Sigma-Aldrich
Carbon Film Supported Copper Hexagonal Mesh Grid
size 400 mesh, box of 50
Synonyme(s) :
CARBON FILM ONLY ON 400 HEX COPPER GRIDS 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Matériaux
carbon film
copper grid
Description
film layer thickness 5 - 6 nm carbon
Conditionnement
box of 50
Taille
400 mesh
Description générale
A 5-6 nm thin film of pure carbon supported hexagonal mesh grids. Copper grid, 400 mesh. Carbon film show better electrical and heat conduction in the electron beam.