Principaux documents
TEM-CF300CUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Copper Square Mesh
size 300 mesh, box of 50
Synonyme(s) :
CARBON SQUARE MESH - CU 300 MESH UL 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
300 mesh
Description générale
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Copper grid, 300 mesh.