Principaux documents
TEM-7596410
Sigma-Aldrich
OMNI Probe Copper 4-Post Lift-Out Grid
pack of 100 (per vial)
Synonyme(s) :
Omni Grids - Copper 4-Post Lift-Out Grids 100/pk
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Le produit TEM-7596410 n'est pas actuellement commercialisé dans votre pays Contacter notre Service technique
Conditionnement
pack of 100 (per vial)
Catégories apparentées
Description générale
4 post copper lift-out grids specifically designed for in-situ lift-out. These grids include multiple indexed mounting locations, two with vertical bar attachment surfaces and two with "V" shaped alighment surfaces. Sides have lower profile for easier access to outermost posts. 3mm diameter.
Conditionnement
Packaged in plastic vials.