Principaux documents
SYNM280004
Synthware all-glass vacuum/inert gas manifold with solid high vacuum stopcocks
number of ports, 4, Hose Connections: Front-left, Rear-left-right
Synonyme(s) :
Synthware M280004, vacuum manifold, vacuum unit
1 EA
1 000,00 $
1 000,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
1 EA
1 000,00 $
About This Item
Code UNSPSC :
40161501
Nomenclature NACRES :
NB.51
1 000,00 $
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Caractéristiques
number of ports 4
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware M280004
Description générale
With 4 mm double oblique, high vacuum solid glass stopcocks precision ground with corresponding numbers on barrel and plug. Solid glass stopcocks have constant pressure retention devices. Beams are 22 mm O.D. with front-left and rear-left-right 10 mm hose connections. Distance between stopcocks is 75 mm center to center.
Informations légales
Synthware is a trademark of Kemtech America Inc.