SYND251014
Synthware™ Vigreux microscale distillation head
joint: ST/NS 14/20, immersion depth 25 mm
Synonyme(s) :
Synthware D251014, short path, distilling head
About This Item
Produits recommandés
Joint
joint: ST/NS14/20
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware D251014
Profondeur d'immersion
25 mm
Description générale
With Vigreux Indentations, used with 25 mm immersion thermometer, and D27 distillation receiver.
Informations légales
Synthware is a trademark of Kemtech America Inc.