Principaux documents
SYNA462424
Synthware modified anti-splash adapter with fritted disc
100 mL, top joint: ST/NS 24/40, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A462424, Anti-splash adapter, splash head, bump trap
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A462424
Capacité
100 mL
Catégories apparentées
Description générale
With extra coarse fritted disc sealed above lower inner joint. Frit reduces foam overflow from being drawn into the condenser.
Informations légales
Synthware is a trademark of Kemtech America Inc.