Principaux documents
SYNA302440
Synthware vacuum or argon/nitrogen adapter with hose connection on top
joint: ST/NS 24/40
Synonyme(s) :
Synthware A302440, inert gas adapter, vacuum adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
joint: ST/NS24/40
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A302440
Catégories apparentées
Description générale
With hose connection on the top and male joint.
Informations légales
Synthware is a trademark of Kemtech America Inc.