Principaux documents
SYNA252924
Synthware enlarging adapter
top joint: ST/NS 29/42, bottom joint: ST/NS 24/40
Synonyme(s) :
Synthware A252924, glass connecting adapter
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.51
Joint
bottom joint: ST/NS24/40
joint: ST/NS top29/42
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
Synthware A252924
Description générale
Synthware enlarging adapter is used for connecting dissimilar standard taper joint sizes. The top joint (29/42) is larger that the bottom joint (24/40).
Informations légales
Synthware is a trademark of Kemtech America Inc.