Documents
SLW1060/20D
Pyrex® Watch glasses
nominal diameter 150 mm
Synonyme(s) :
Pyrex, SciLabware 1060/20D
About This Item
Code UNSPSC :
41121801
Nomenclature NACRES :
NB.43
Conditionnement
pkg of 10
Fabricant/nom de marque
1060/20D
SciLabware
Diamètre nominal
150 mm
Description générale
Watch glasses
  • For evaporation work and covering heated beakers
  • Manufactured from heat resisting Pyrex borosilicate glass
  • Fire-polished edges to reduce chipping and breakage
Informations légales
PYREX is a registered trademark of Corning, Inc.