Principaux documents
MT59902371
Mettler-Toledo electrode cable
L 1.2 m
About This Item
Fabricant/nom de marque
Mettler-Toledo 59902371
Longueur
1.2 m
Description générale
Mettler Toledo electrodes can easily be connected to various third-party instruments. All you have to do is select the appropriate cable. We offer a wide choice of cable lengths and plug combinations. For Mettler Toledo reference electrodes and temperature probes.