Principaux documents
MT51107672
Mettler Toledo DX series ISE half-cells electrode
Cadmium selective
About This Item
Code UNSPSC :
39121436
Nomenclature NACRES :
NB.79
Matériaux
membrane (polymer)
shaft (POM/PVC)
Fabricant/nom de marque
Mettler-Toledo 51107672
Plage de mesure
1.0-1.0-3 mmol/L
Plage de mesure de la température
0-50 °C
pH
2-8
Description générale
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 1 mol/L KNO3