Principaux documents
MDFNSPF-L
Mediflex Nisense Gloves, blue nitrile, powder free
size L
About This Item
Code UNSPSC :
46181504
Matériaux
sky blue nitrile
Taille
L
Description
textured
Caractéristiques
latex free
powder-free
Conditionnement
box of 100 gloves
Longueur
240 mm , cuff
Description générale
Powder free blue nitrile gloves. Non sterile, ambidextrous, latex
free, ultra thin, 240mm cuff, textured. Single use only.