Documents
ESYNTH001
Sigma-Aldrich
SynLectro 50 mL starter kit
Synonyme(s) :
SynLectro 50 mL Starter Set
About This Item
Code UNSPSC :
81141502
Nomenclature NACRES :
NB.61
Description générale
SynLectro 50 mL Starter Kit is the perfect way to get your electrochemical reaction started.
This kit contains:
  • 50 mL glass cell
  • PTFE stopper and sleeve
  • 2 electrode holders
  • 2 graphite electrodes
  • stainless steel electrode
  • nickel electrode
  • lead electrode
  • zinc electrode
SynLectro Electrolysis Platform
The SynLectro Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Informations légales
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Contenu apparenté
Learn about our new SynLectro™ Electrolysis Platform that has been designed to simplify and standardize electrochemical organic synthesis at research and batch scale.