Documents
DWK241701401
DURAN® reaction flask with ground joint
glass flask, flask capacity (25 mL), single neck, round bottom, neck joint: ST/NS 19/26
Synonyme(s) :
241701401
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
glass flask
round bottom
Joint
joint: ST/NS neck19/26
Caractéristiques
lid
Conditionnement
case of 10 ea
Fabricant/nom de marque
DURAN® 241701401
Contenance du flacon/de la fiole
(25 mL)
Description générale
Tight closure due to standard ground joint. The geometry permits very uniform heating. Closable with glass and plastic stopper
Informations légales
DURAN is a registered trademark of DWK Life Sciences